missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-N-[3-(5-methyl-1,2,4-oxadiazol-3-yl)benzyl]amine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 250 mg |
| Formula Weight | 203.24 |
| Percent Purity | 97% |
| Chemical Name or Material | N-methyl-N-[3-(5-methyl-1, 2, 4-oxadiazol-3-yl)benzyl]amine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction