missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(5-methyl-2-phenyl-3-furyl)methyl]amine, 95%, Thermo Scientific™

Product Code. 10537484
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10537484 1 g 1g
11330677 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10537484 Supplier Thermo Scientific Maybridge Supplier No. CC49746DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 869901-18-8
Molecular Formula C13H15NO
Molecular Weight (g/mol) 201.27
MDL Number MFCD08690282
InChI Key UOPUGXOLGTYISK-UHFFFAOYSA-N
Synonym n-methyl-n-5-methyl-2-phenyl-3-furyl methyl amine, 3-furanmethanamine,n,5-dimethyl-2-phenyl, methyl 5-methyl-2-phenylfuran-3-yl methyl amine, methyl 5-methyl-2-phenyl 3-furyl methyl amine, n-methyl-1-5-methyl-2-phenyl-3-furyl methylamine, n-methyl-1-5-methyl-2-phenylfuran-3-yl methanamine
PubChem CID 18525837
IUPAC Name N-methyl-1-(5-methyl-2-phenylfuran-3-yl)methanamine
SMILES CNCC1=C(OC(C)=C1)C1=CC=CC=C1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Quantity 1 g
Formula Weight 201.27
Percent Purity 90%
Chemical Name or Material N-methyl-N-[(5-methyl-2-phenyl-3-furyl)methyl]amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.