missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-propargylbenzylamine, 97%

Product Code. 10193220 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10193220

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 555-57-7 | C11H13N | 159.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-57-7
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.23
MDL Number MFCD00008576
InChI Key DPWPWRLQFGFJFI-UHFFFAOYSA-N
Synonym pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
PubChem CID 4688
ChEBI CHEBI:7930
IUPAC Name N-benzyl-N-methylprop-2-yn-1-amine
SMILES CN(CC#C)CC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9400g/mL
Boiling Point 86°C to 88°C (4.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH2N(CH3)CH2C∽CH
Refractive Index 1.5213
Quantity 5 g
Beilstein 12, II, 548
Merck Index 15, 7142
Specific Gravity 0.94
Solubility Information Solubility in water: soluble. Other solubilities: miscibel in ethanol,acetone
Formula Weight 159.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Methyl-N-propargylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Methyl-N-propargylbenzylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.