missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-p-anisidine, 98%

Product Code. 10265990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265990 10 g Glass bottle 10g
10174190 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10265990 Supplier Thermo Scientific Acros Supplier No. 177220100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5961-59-1 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5961-59-1
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008399
InChI Key JFXDIXYFXDOZIT-UHFFFAOYSA-N
Synonym n-methyl-p-anisidine, n-methyl-4-anisidine, n-methyl-4-methoxyaniline, benzenamine, 4-methoxy-n-methyl, n-methyl-p-ansidine, p-anisidine, n-methyl, 4-methoxy-n-methylbenzenamine, 1-methylamino-4-methoxybenzene, p-methoxy-n-methylaniline, acmc-209mes
PubChem CID 22250
IUPAC Name 4-methoxy-N-methylaniline
SMILES CNC1=CC=C(C=C1)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33.0°C to 36.0°C
Color Brown-Gray to White
Boiling Point 135.0°C to 136.0°C (19.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H4NHCH3
Quantity 10 g
Beilstein 13, 442
Formula Weight 137.18
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material N-Methyl-p-anisidine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methyl-p-anisidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.