missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl Rivastigmine Chloride, TRC

Product Code. 18044040
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18044040 100 mg 100mg
18023840 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18044040 Supplier Toronto Research Chemicals Supplier No. TRCM326340100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15 H25 N2 O2 . Cl
Molecular Weight (g/mol) 300.82
Synonym (S)-1-(3-{[Ethyl(methyl)carbamoyl]oxy}phenyl)-N,N,N-trimethylethanaminium chloride, Rivastigmine Related Compound E
IUPAC Name [(1S)-1-[3-[ethyl(methyl)carbamoyl]oxyphenyl]ethyl]-trimethylazanium;chloride
SMILES [Cl-].CCN(C)C(=O)Oc1cccc(c1)[C@H](C)[N+](C)(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H25N2O2.ClH/c1-7-16(3)15(18)19-14-10-8-9-13(11-14)12(2)17(4,5)6;/h8-12H,7H2,1-6H3;1H/q+1;/p-1/t12-;/m0./s1
Formula Weight 300.1605
Chemical Name or Material (S)-1-[3-[(Ethyl(methyl)carbamoyl)oxy]phenyl]-N,N,N-trimethylethanaminium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.