missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(tetrahydrothiopyran-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10493034
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10493034 250 mg Amber Glass Bottle 250mg
10664785 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10493034 Supplier Thermo Scientific Maybridge Supplier No. CC61946CB
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 950603-22-2
Molecular Formula C7H15NS
Molecular Weight (g/mol) 145.264
MDL Number MFCD09879924
InChI Key FPKJBESBCAZWPI-UHFFFAOYSA-N
Synonym methyl thian-4-ylmethyl amine, n-methyl tetrahydrothiopyran-4-yl methylamine, n-methyl tetrahydro-2h-thiopyran-4-yl methylamine, methyl thian-4-yl methyl amine, n-methyl-1-thian-4-yl methanamine, n-methyl-n-tetrahydro-2h-thiopyran-4-ylmethyl amine
PubChem CID 24229643
IUPAC Name N-methyl-1-(thian-4-yl)methanamine
SMILES CNCC1CCSCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 145.27
Percent Purity 97%
Chemical Name or Material N-Methyl(tetrahydrothiopyran-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.