missing translation for 'onlineSavingsMsg'
Learn More

N-Methylanthranilic acid, 90+%

Product Code. 11438730
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438730 25 g 25g
11468897 100 g 100g
11478897 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438730 Supplier Thermo Scientific Alfa Aesar Supplier No. B20531.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-68-6 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-68-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002424
InChI Key WVMBPWMAQDVZCM-UHFFFAOYSA-N
Synonym n-methylanthranilic acid, 2-methylamino benzoic acid, n-methyl-2-aminobenzoic acid, n-methyl-o-aminobenzoic acid, benzoic acid, 2-methylamino, o-methylamino benzoic acid, anthranilic acid, n-methyl, 2-methylamino-benzoic acid, kyselina n-methylanthranilova, unii-vpb2514iuj
PubChem CID 67069
ChEBI CHEBI:16394
IUPAC Name 2-(methylamino)benzoic acid
SMILES CNC1=CC=CC=C1C(=O)O

Specifications

Melting Point ∼171°C (decomposition)
Quantity 25 g
Beilstein 2208135
Formula Weight 151.17
Percent Purity ≥90%
Chemical Name or Material N-Methylanthranilic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.