missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbenzylamine, 97%

Product Code. 10090590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
Unit Size:
500mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10090590 5 mL Glass bottle 5mL
10419400 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10090590 Supplier Thermo Scientific Acros Supplier No. 106040050
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 103-67-3 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-67-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008289
InChI Key RIWRFSMVIUAEBX-UHFFFAOYSA-N
Synonym n-methylbenzylamine, methylbenzylamine, n-benzylmethylamine, benzyl methyl amine, benzenemethanamine, n-methyl, benzylmethylamine, n-methy-n-benzylamine, n-benzyl-n-methylamine, benzylamine, n-methyl, n-methyl phenyl methanamine
PubChem CID 7669
IUPAC Name N-methyl-1-phenylmethanamine
SMILES CNCC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -24°C
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 184°C to 189°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH2NHCH3
Refractive Index 1.5214 to 1.5234
Quantity 5 mL
Beilstein 12, 1019
Specific Gravity 0.94
Solubility Information Solubility in water: 65g/L (20°C)
Formula Weight 121.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Methylbenzylamine, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.