missing translation for 'onlineSavingsMsg'
Learn More

N-Methylhippuric acid, 99%, Thermo Scientific™

Product Code. p-164451 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10710561

Brand: Thermo Scientific Acros 259760050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2568-34-5
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00144940
InChI Key PKCSYDDSNIJRIX-UHFFFAOYSA-N
Synonym benzoylsarcosine, n-methylhippuric acid, n-benzoyl-n-methylglycine, n-benzoylsarcosine, glycine, n-benzoyl-n-methyl, 2-n-methylbenzamido acetic acid, sarcosine, n-benzoyl, 2-benzoyl methyl amino acetic acid, chembl66391, 2-n-methyl-1-phenylformamido acetic acid
PubChem CID 75728
IUPAC Name 2-[benzoyl(methyl)amino]acetic acid
SMILES CN(CC(O)=O)C(=O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 101°C to 105°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5C(O)N(CH3)CH2COOH
Quantity 5 g
Formula Weight 193.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Methylhippuric acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methylhippuric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.