missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzylethylenediamine, 97%, ACROS Organics™

Product Code. 10011704 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10011704 25 g Glass bottle 25g
10525724 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10011704 Supplier Thermo Scientific Acros Supplier No. 112620250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 140-28-3
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00004771
InChI Key JUHORIMYRDESRB-UHFFFAOYSA-N
Synonym n,n'-dibenzylethylenediamine, benzathine, dbed, benzatin, n1,n2-dibenzylethane-1,2-diamine, 1,2-bis benzylamino ethane, 1,2-ethanediamine, n,n'-bis phenylmethyl, usaf do-53, unii-c659vz7p7t, ethylenediamine, n,n'-dibenzyl
PubChem CID 8793
ChEBI CHEBI:51344
IUPAC Name N,N'-dibenzylethane-1,2-diamine
SMILES C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.024g/mL
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH2NHCH2CH2NHCH2C6H5
Quantity 25 g
Beilstein 12, 1067
Merck Index 15, 1066
Formula Weight 240.35
Chemical Name or Material N, N'-Dibenzylethylenediamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.