missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzylethylenediamine, 97%

Product Code. 11362206
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362206 100 g 100g
11352206 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362206 Supplier Thermo Scientific Alfa Aesar Supplier No. L03540.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 140-28-3 | C16H20N2 | 240.35 g/mol

N,N'-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-28-3
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00004771
InChI Key JUHORIMYRDESRB-UHFFFAOYSA-N
Synonym n,n'-dibenzylethylenediamine, benzathine, dbed, benzatin, n1,n2-dibenzylethane-1,2-diamine, 1,2-bis benzylamino ethane, 1,2-ethanediamine, n,n'-bis phenylmethyl, usaf do-53, unii-c659vz7p7t, ethylenediamine, n,n'-dibenzyl
PubChem CID 8793
ChEBI CHEBI:51344
IUPAC Name N,N'-dibenzylethane-1,2-diamine
SMILES C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2

Specifications

Melting Point 23°C to 25°C
Density 1.025
Boiling Point 184°C to 186°C (2 mmHg)
Flash Point >110°C (230°F)
Linear Formula C6H5CH2NHCH2CH2NHCH2C6H5
Refractive Index 1.565
Quantity 100 g
UN Number UN2735
Beilstein 786668
Sensitivity Air Sensitive
Merck Index 14,1064
Solubility Information Slightly soluble in water.
Formula Weight 240.35
Percent Purity 97%
Chemical Name or Material N,N'-Dibenzylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.