missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzylethylenediamine diacetate, 99%

Product Code. 11324865
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324865 500 g 500g
11304865 25 g 25g
11314865 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324865 Supplier Thermo Scientific Alfa Aesar Supplier No. L17974.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-75-8 | C20H28N2O4 | 360.454 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-75-8
Molecular Formula C20H28N2O4
Molecular Weight (g/mol) 360.454
MDL Number MFCD00040588
InChI Key MTRNNCLQPVCDLF-UHFFFAOYSA-N
Synonym n,n'-dibenzylethylenediamine diacetate, n1,n2-dibenzylethane-1,2-diamine diacetate, n,n'-dibenzyl ethylenediamine diacetate, benzathine diacetate, dbed diacetate, unii-x18b8x2c22, n,n'-dibenzylethylenediammonium di acetate, 1,2-ethanediamine, n,n'-bis phenylmethyl-, diacetate, bis acetic acid ; benzathine, 1,2-di benzylamino ethane diacetate
PubChem CID 31228
IUPAC Name acetic acid;N,N'-dibenzylethane-1,2-diamine
SMILES CC(=O)O.CC(=O)O.C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2

Specifications

Melting Point 117°C to 118°C
Quantity 500 g
Beilstein 3781362
Merck Index 14,1064
Formula Weight 360.46
Percent Purity 99%
Chemical Name or Material N,N'-Dibenzylethylenediamine diacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.