missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzylthiourea, 98%, Thermo Scientific™

Product Code. 11361567
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361567 25 g 25g
11371567 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361567 Supplier Thermo Scientific Alfa Aesar Supplier No. L04390.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1424-14-2
Molecular Formula C15H16N2S
Molecular Weight (g/mol) 256.367
MDL Number MFCD00022013
InChI Key LQZPSWMMTICWHD-UHFFFAOYSA-N
Synonym n,n'-dibenzylthiourea, sym-dibenzylthiourea, thiourea, n,n'-bis phenylmethyl, 1,3-dibenzyl-2-thiourea, thiourea, bis phenylmethyl, urea, 1,3-dibenzyl-2-thio, dibenzylthiourea, acmc-20ak9g, n,n/'-dibenzylthiourea, urea,3-dibenzyl-2-thio
PubChem CID 838375
IUPAC Name 1,3-dibenzylthiourea
SMILES C1=CC=C(C=C1)CNC(=S)NCC2=CC=CC=C2

Specifications

Melting Point 146°C to 149°C
Odor Odorless
Quantity 25 g
Beilstein 989212
Formula Weight 256.37
Percent Purity 98%
Chemical Name or Material N,N'-Dibenzylthiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.