missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylhydroxylamine, 97%

Product Code. 11440907
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440907 100 g 100g
11430907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440907 Supplier Thermo Scientific Alfa Aesar Supplier No. B23302.22
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3710-84-7 | C4H11NO | 89.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3710-84-7
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00002126
InChI Key FVCOIAYSJZGECG-UHFFFAOYSA-N
Synonym diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamine
PubChem CID 19463
IUPAC Name N,N-diethylhydroxylamine
SMILES CCN(CC)O

Specifications

Melting Point -25°C
Density 0.867
Boiling Point 125°C to 130°C
Flash Point 46°C (114°F)
Refractive Index 1.4215
Quantity 100 g
UN Number UN1993
Beilstein 1731349
Sensitivity Hygroscopic
Formula Weight 89.14
Percent Purity 97%
Chemical Name or Material N,N-Diethylhydroxylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Diethylhydroxylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: N,N-Diethylhydroxylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.