missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbenzylamine, 99%

Product Code. 10156504 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
100mL
2.5 L
5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10156504 2.5 L Glass bottle 2.5 L
10635022 100 mL Glass bottle 100mL
10531411 500 mL Glass bottle 500mL
10564014 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10156504 Supplier Thermo Scientific Acros Supplier No. 159750025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-83-3 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-83-3
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008329
InChI Key XXBDWLFCJWSEKW-UHFFFAOYSA-N
Synonym n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
PubChem CID 7681
SMILES CN(C)CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -75.0°C
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 183.0°C to 184.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2N(CH3)2
Refractive Index 1.5000 to 1.5020
Quantity 2.5 L
Beilstein 12, 1019
Specific Gravity 0.9
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: soluble in alcohol,acetone,ether and toluene,reacts with organic and inorganic acids
IUPAC Name N,N-dimethyl-1-phenylmethanamine
Formula Weight 135.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylbenzylamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.