missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide dibenzyl acetal, tech. 80%

Product Code. 11433644
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433644 10 g 10g
11443644 50 g 50g
11453644 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433644 Supplier Thermo Scientific Alfa Aesar Supplier No. A11281.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2016-04-8 | C17H22NO2 | 272.37 g/mol

N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.

Solubility
Soluble in variety of inert solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2016-04-8
Molecular Formula C17H22NO2
Molecular Weight (g/mol) 272.37
MDL Number MFCD00014869
InChI Key JFIKHFNGAURIIB-UHFFFAOYSA-O
Synonym n,n-dimethylformamide dibenzyl acetal, n,n-dimethylformamide dibenzylacetal, 1,1-bis benzyloxy trimethylamine, methanamine, n,n-dimethyl-1,1-bis phenylmethoxy, bis benzyloxy methyl dimethylamine, acmc-1cro2, 1,1-dibenzyloxytrimethylamine, dimethyl formamide dibenzylacetal, bis benzyloxy methyldimethylamine, dimethyl formamide dibenzyl acetal
PubChem CID 74837
IUPAC Name N,N-dimethyl-1,1-bis(phenylmethoxy)methanamine
SMILES CN(C)C(OCC1=CC=CC=C1)OCC2=CC=CC=C2

Specifications

Density 1.045
Boiling Point 138°C to 140°C (0.5 mmHg)
Quantity 10 g
Sensitivity Moisture sensitive
Solubility Information Soluble in variety of inert solvents.
Formula Weight 271.36
Percent Purity 85%
Grade Technical
Chemical Name or Material N,N-Dimethylformamide dibenzyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.