missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N-desmethyl-diltiazem, TRC

Product Code. 30464382
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464382 25 mg 25mg
30469475 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30464382 Supplier Toronto Research Chemicals Supplier No. TRCN27733025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso-desmethyl-diltiazem
Analyte or Component Names N-Nitroso-desmethyl-diltiazem
Shipping Condition Room Temperature
Molecular Formula C21H23N3O5S
Synonym (2S,3S)-2-(4-Methoxyphenyl)-5-(2-(methyl(nitroso)amino)ethyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl Acetate
InChI Formula InChI=1S/C21H23N3O5S/c1-14(25)29-19-20(15-8-10-16(28-3)11-9-15)30-18-7-5-4-6-17(18)24(21(19)26)13-12-23(2)22-27/h4-11,19-20H,12-13H2,1-3H3/t19-,20+/m1/s1
Solution Type Neat
SMILES CC(O[C@@H]1[C@@H](SC2=CC=CC=C2N(CCN(N=O)C)C1=O)C3=CC=C(OC)C=C3)=O
IUPAC Name (2S,3S)-2-(4-methoxyphenyl)-5-(2-(methyl(nitroso)amino)ethyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 429.49
Quantity 25 mg
Formula Weight 429.1358
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.