missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosometoprolol, TRC

Product Code. 30007501
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007501 50 mg 50mg
17925075 25 mg 25mg
17980435 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007501 Supplier Toronto Research Chemicals Supplier No. TRCN13729050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138768-62-4
Molecular Formula C15H24N2O4
Molecular Weight (g/mol) 296.36
Synonym 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)nitrosoamino]-, 1-[4-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)nitrosoamino]-2-propanol
IUPAC Name N-[2-hydroxy-3-[4-(2-methoxyethyl)phenoxy]propyl]-N-propan-2-ylnitrous amide
SMILES COCCc1ccc(OCC(O)CN(N=O)C(C)C)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H24N2O4/c1-12(2)17(16-19)10-14(18)11-21-15-6-4-13(5-7-15)8-9-20-3/h4-7,12,14,18H,8-11H2,1-3H3
Formula Weight 296.1736
Percent Purity >95%
Chemical Name or Material N-Nitrosometoprolol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.