missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrososotalol, TRC

Product Code. 17981685
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17981685 250 mg 250mg
17901775 50 mg 50mg
17954265 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17981685 Supplier Toronto Research Chemicals Supplier No. TRCN137340250MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134720-07-3
Molecular Formula C12 H19 N3 O4 S
Molecular Weight (g/mol) 301.362
Synonym Methanesulfonamide, N-[4-[1-hydroxy-2-[(1-methylethyl)nitrosoamino]ethyl]phenyl]-, N-[4-[1-Hydroxy-2-[(1-methylethyl)nitrosoamino]ethyl]phenyl]methanesulfonamide, N-Nitrososotalol;Methanesulfonamide, N-[4-[1-hydroxy-2-[(1-methylethyl)nitrosoamino]ethyl]phenyl]-, N-[4-[1-Hydroxy-2-[(1-methylethyl)nitrosoamino]ethyl]phenyl]methanesulfonamide
IUPAC Name N-[4-[1-hydroxy-2-[nitroso(propan-2-yl)amino]ethyl]phenyl]methanesulfonamide
SMILES CC(C)N(CC(O)c1ccc(NS(=O)(=O)C)cc1)N=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H19N3O4S/c1-9(2)15(14-17)8-12(16)10-4-6-11(7-5-10)13-20(3,18)19/h4-7,9,12-13,16H,8H2,1-3H3
Formula Weight 301.11
Percent Purity >95%
Chemical Name or Material N-Nitrososotalol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.