missing translation for 'onlineSavingsMsg'
Learn More

(+)-N,O-Didesmethyl Tramadol, TRC

Product Code. 30372648
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372648 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372648 Supplier Toronto Research Chemicals Supplier No. TRCD4412305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 144830-18-2
Molecular Formula C14 H21 N O2
Quantity 5 mg
Synonym Phenol, 3-[(1R,2R)-1-hydroxy-2-[(methylamino)methyl]cyclohexyl]-, Phenol, 3-[1-hydroxy-2-[(methylamino)methyl]cyclohexyl]-, (1R-cis)-, 3-[(1R,2R)-1-Hydroxy-2-[(methylamino)methyl]cyclohexyl]phenol, (+)-N,O-Didesmethyl Tramadol, (+)-N,O-Didesmethyltramadol, (R,R)-N,O-Didesmethyltramadol, 3-[(1R-cis)-1-Hydroxy-2-[(methylamino)methyl]cyclohexyl]phenol
InChI Formula InChI=1S/C14H21NO2/c1-15-10-12-5-2-3-8-14(12,17)11-6-4-7-13(16)9-11/h4,6-7,9,12,15-17H,2-3,5,8,10H2,1H3/t12-,14+/m1/s1
SMILES CNC[C@H]1CCCC[C@]1(O)c2cccc(O)c2
IUPAC Name 3-[(1R,2R)-1-hydroxy-2-(methylaminomethyl)cyclohexyl]phenol
Molecular Weight (g/mol) 235.32
Formula Weight 235.1572
Chemical Name or Material (+)-N,O-Di-Desmethyl Tramadol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.