missing translation for 'onlineSavingsMsg'
Learn More

N-(o-Tolyl)thiourea, 97+%, Thermo Scientific™

Product Code. 11486066
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486066 250 g 250g
11466066 10 g 10g
11476066 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486066 Supplier Thermo Scientific Alfa Aesar Supplier No. B22350.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-78-8
Molecular Formula C8H10N2S
Molecular Weight (g/mol) 166.24
MDL Number MFCD00041162
InChI Key ACLZYRNSDLQOIA-UHFFFAOYSA-N
Synonym o-tolylthiourea, 2-methylphenyl thiourea, n-o-tolyl thiourea, 1-2-methylphenyl thiourea, n-2-methylphenyl thiourea, 1-2-methylphenyl-2-thiourea, 2-tolylthiourea, 1-o-tolyl-2-thiourea, thiourea, 2-methylphenyl, 1-o-tolyl thiourea
PubChem CID 2787703
IUPAC Name (2-methylphenyl)thiourea
SMILES CC1=CC=CC=C1NC(N)=S

Specifications

Melting Point 155°C to 158°C
Odor Odorless
Quantity 250 g
UN Number UN2811
Beilstein 2086802
Formula Weight 166.25
Percent Purity ≥97%
Chemical Name or Material N-(o-Tolyl)thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.