missing translation for 'onlineSavingsMsg'
Learn More

n-Pentyl 4-hydroxybenzoate, 98%

Product Code. 11357495
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357495 5 g 5g
11367495 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357495 Supplier Thermo Scientific Alfa Aesar Supplier No. L05563.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6521-29-5 | C12H16O3 | 208.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
n-Pentyl 4-hydroxybenzoate, is used as an antimicrobial agent in cosmetic products.

Solubility
Insoluble in water. Slightly soluble in chloroform, methanol.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6521-29-5
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00016479
InChI Key ZNSSPLQZSUWFJT-UHFFFAOYSA-N
PubChem CID 23019
IUPAC Name pentyl 4-hydroxybenzoate
SMILES CCCCCOC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 36°C to 40°C
Boiling Point 197°C to 200°C (10 mmHg)
Quantity 5 g
Beilstein 2722438
Synonym amyl 4-hydroxybenzoate, pentyl p-hydroxybenzoate, pentylparaben, n-pentyl 4-hydroxybenzoate, benzoic acid, 4-hydroxy-, pentyl ester, p-hydroxybenzoic acid pentyl ester, pentyl paraben, unii-sjv2e4c7dm, p-oxybenzoesaurepentylester, amyl4-hydroxybenzoate
Solubility Information Insoluble in water. Slightly soluble in chloroform,methanol.
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material n-Pentyl 4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.