missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Pentyl 4-hydroxybenzoate, 98%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
n-Pentyl 4-hydroxybenzoate, is used as an antimicrobial agent in cosmetic products.
Solubility
Insoluble in water. Slightly soluble in chloroform, methanol.
Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
n-Pentyl 4-hydroxybenzoate, is used as an antimicrobial agent in cosmetic products.
Solubility
Insoluble in water. Slightly soluble in chloroform, methanol.
Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | 36°C to 40°C |
| Boiling Point | 197°C to 200°C (10 mmHg) |
| Quantity | 5 g |
| Beilstein | 2722438 |
| Synonym | amyl 4-hydroxybenzoate, pentyl p-hydroxybenzoate, pentylparaben, n-pentyl 4-hydroxybenzoate, benzoic acid, 4-hydroxy-, pentyl ester, p-hydroxybenzoic acid pentyl ester, pentyl paraben, unii-sjv2e4c7dm, p-oxybenzoesaurepentylester, amyl4-hydroxybenzoate |
| Solubility Information | Insoluble in water. Slightly soluble in chloroform,methanol. |
| Formula Weight | 208.26 |
| Percent Purity | 98% |
| Chemical Name or Material | n-Pentyl 4-hydroxybenzoate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction