missing translation for 'onlineSavingsMsg'
Learn More

n-Pentyl benzoate, 98+%

Product Code. 11449472
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449472 100 g 100g
11459472 500 g 500g
11469472 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449472 Supplier Thermo Scientific Alfa Aesar Supplier No. A10177.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2049-96-9 | C12H16O2 | 192.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2049-96-9
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00048843
InChI Key QKNZNUNCDJZTCH-UHFFFAOYSA-N
Synonym amyl benzoate, n-pentyl benzoate, benzoic acid, pentyl ester, n-amyl benzoate, unii-cle7k4b29s, cle7k4b29s, amylbenzoat, amylbenzoate, n amyl benxoate, benzoic acid pentyl
PubChem CID 16296
IUPAC Name pentyl benzoate
SMILES CCCCCOC(=O)C1=CC=CC=C1

Specifications

Density 0.99
Boiling Point 260°C
Flash Point 130°C (266°F)
Refractive Index 1.495
Quantity 100 g
Formula Weight 192.26
Percent Purity ≥98%
Chemical Name or Material n-Pentyl benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.