missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-3-(trifluoromethyl)aniline, TRC

Product Code. 30136085
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136085 100 mg 100mg
30135210 500 mg 500mg
30004210 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136085 Supplier Toronto Research Chemicals Supplier No. TRCP399710100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-23-5
Molecular Formula C13 H10 F3 N
Molecular Weight (g/mol) 237.22
Synonym N-Phenyl-3-(trifluoromethyl)aniline, Etofenamate Imp. C (EP)
IUPAC Name N-phenyl-3-(trifluoromethyl)aniline
SMILES FC(F)(F)c1cccc(Nc2ccccc2)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H10F3N/c14-13(15,16)10-5-4-8-12(9-10)17-11-6-2-1-3-7-11/h1-9,17H
Formula Weight 237.0765
Chemical Name or Material N-Phenyl-3-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.