missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylpropanamide, TRC

Product Code. 30418074
Change view
Click to view available options
Quantity:
50 mg
1 g
2.5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418074 50 mg -
30408850 2.5 g 2.5g
30417455 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418074 Supplier Toronto Research Chemicals Supplier No. TRCP33606050MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-71-3
Molecular Formula C9 H11 N O
Molecular Weight (g/mol) 149.19
Synonym N-Phenylpropanamide, Fentanyl Imp. G (EP), Propanamide, N-phenyl-, Propionanilide (6CI,7CI,8CI), N-Phenylpropionamide, NSC 58952, R 50977
IUPAC Name N-phenylpropanamide
SMILES CCC(=O)Nc1ccccc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H11NO/c1-2-9(11)10-8-6-4-3-5-7-8/h3-7H,2H2,1H3,(H,10,11)
Formula Weight 149.0841
Percent Purity >95%
Chemical Name or Material N-Phenylpropanamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.