accessibility menu, dialog, popup

UserName

CAS RN 620-71-3

IUPAC Name: N-phenylpropanamide
Synonyms: propionanilide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: ZTHRQJQJODGZHV-UHFFFAOYSA-N
SMILES: CCC(=O)NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results