missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylurethane, 98%

Product Code. 11341797
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341797 25 g 25g
11351797 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341797 Supplier Thermo Scientific Alfa Aesar Supplier No. L05027.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 101-99-5 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-99-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00026806
InChI Key LBKPGNUOUPTQKA-UHFFFAOYSA-N
Synonym n-phenylurethane, phenylurethane, ethyl phenylcarbamate, phenylurethan, ethyl carbanilate, euphorin, keimstop, urethane, phenyl, carbanilic acid, ethyl ester, phenylethyl carbamate
PubChem CID 7591
IUPAC Name ethyl N-phenylcarbamate
SMILES CCOC(=O)NC1=CC=CC=C1

Specifications

Melting Point 49°C to 51°C
Density 1.106
Boiling Point 237°C
Refractive Index 1.5375
Quantity 25 g
Beilstein 1942785
Merck Index 14,7320
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material N-Phenylurethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.