missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl 4-hydroxybenzoate, 99+%

Product Code. 11429482
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429482 5000 g 5000g
11419482 250 g 250g
11409482 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429482 Supplier Thermo Scientific Alfa Aesar Supplier No. A10180.0I
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 94-13-3 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
n-Propyl 4-hydroxybenzoate is used as antimicrobial preservatives in pharmaceuticals, foods, cosmetics and shampoos. It is widely used as preservatives in personal care products like creams, lotions and bath products. Further, it is used as a food additive. Its derivative, sodium propyl p-hydroxybenzoate is used as an anti-fungal preservation agent.

Solubility
Soluble in propylene glycol, ethanol, ether, acetone, methanol anddimethyl sulfoxide. Slightly soluble in glycerin. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-13-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00002354
InChI Key QELSKZZBTMNZEB-UHFFFAOYSA-N
Synonym propylparaben, propyl paraben, 4-hydroxybenzoic acid propyl ester, propyl p-hydroxybenzoate, nipasol, nipazol, propyl butex, betacide p, propyl parahydroxybenzoate, propylparasept
PubChem CID 7175
ChEBI CHEBI:32063
IUPAC Name propyl 4-hydroxybenzoate
SMILES CCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 95°C to 99°C
Flash Point 180°C (356°F)
Quantity 5000 g
Beilstein 1103245
Merck Index 14,7866
Solubility Information Soluble in propylene glycol,ethanol,ether,acetone,methanol anddimethyl sulfoxide. Slightly soluble in glycerin. Insoluble in water.
Formula Weight 180.2
Percent Purity ≥99%
Chemical Name or Material n-Propyl 4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.