missing translation for 'onlineSavingsMsg'
Learn More

N-Propylphosphorothioic Triamide, TRC

Product Code. 30011646
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011646 1 g 1g
30008185 10 mg 10mg
30009046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011646 Supplier Toronto Research Chemicals Supplier No. TRCP8337051G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916809-14-8
Molecular Formula C3 H12 N3 P S
Molecular Weight (g/mol) 153.1862
Synonym N-Propylphosphorothioic Triamide, N-n-Propylthiophosphoric triamide
IUPAC Name N-diaminophosphinothioylpropan-1-amine
SMILES CCCNP(=S)(N)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H12N3PS/c1-2-3-6-7(4,5)8/h2-3H2,1H3,(H5,4,5,6,8)
Formula Weight 153.049
Chemical Name or Material N-Propylphosphorothioic Triamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.