missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-4-methylbenzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15441278
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441278 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15441278 Supplier Thermo Scientific Alfa Aesar Supplier No. H55126.MB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 2849-81-2
Molecular Formula C11H17NO2S
MDL Number MFCD00026270
Quantity 25 mg
Synonym n-tert-butyl-p-toluenesulfonamide, p-toluenesulfonamide, n-tert-butyl, benzenesulfonamide, n-1,1-dimethylethyl-4-methyl, n-tert-butyl-4-methylbenzenesulfonamide, maybridge3_004443, cambridge id 5116324, rarechem am uh v184, n-t-butyl-p-toluenesulfonamide, n-tert-butyl-para-toluenesulfonamide
Solubility Information Soluble in water.
InChI Key NYEPSLKMENGNDO-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(C)(C)C
IUPAC Name N-tert-butyl-4-methylbenzenesulfonamide
Molecular Weight (g/mol) 227.32
PubChem CID 246472
Formula Weight 227.32
Percent Purity 97%
Chemical Name or Material N-tert-Butyl-4-methylbenzenesulfonamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.