missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-Amino-2-methylphenyl)acetamide, 90%, Thermo Scientific™

Product Code. p-2704872
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10344222

Brand: Thermo Scientific Maybridge BTB06485DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 56891-59-9
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.208
MDL Number MFCD00276633
InChI Key GWFPMSIIVJMYRZ-UHFFFAOYSA-N
Synonym n-4-amino-2-methylphenyl acetamide, n1-4-amino-2-methylphenyl acetamide, 4'-amino-2'-methylacetanilide, n-4-amino-2-methyl-phenyl-acetamide, 4-acetamido-m-toluidine, 4-acetylamino-m-toluidine, 4-acetamido-3-methylaniline, 4-acetamino-3-methylaniline, acetamide, n-4-amino-2-methylphenyl
PubChem CID 314338
IUPAC Name N-(4-amino-2-methylphenyl)acetamide
SMILES CC1=C(C=CC(=C1)N)NC(=O)C

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 164.21
Percent Purity 90%
Chemical Name or Material N1-(4-amino-2-methylphenyl)acetamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.