missing translation for 'onlineSavingsMsg'
Learn More

N1-[4-Cyano-2-(trifluoromethoxy)phenyl]acetamide, ≥95%, Thermo Scientific™

Product Code. 11366965
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11366965 25 g 25g
10180892 1 g 1g
10233622 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11366965 Supplier Thermo Scientific Maybridge Supplier No. TL00674EB
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-19-0
Molecular Formula C10H7F3N2O2
Molecular Weight (g/mol) 244.173
MDL Number MFCD00204174
InChI Key RHBNAXXJVYFEEA-UHFFFAOYSA-N
Synonym n-4-cyano-2-trifluoromethoxy phenyl acetamide, 4-cyano-2-trifluoromethoxy acetanilide, n1-4-cyano-2-trifluoromethoxy phenyl acetamide, 4'-cyano-2'-trifluoromethoxy acetanilide, maybridge1_000083, 4'-cyano-2'-trifluoromethoxy acetanilid, n-4-cyano-2-trifluoromethoxyphenyl-acetamide
PubChem CID 2736739
IUPAC Name N-[4-cyano-2-(trifluoromethoxy)phenyl]acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)C#N)OC(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%+
Quantity 25 g
Formula Weight 244.17
Percent Purity 95%
Chemical Name or Material N1-[4-Cyano-2-(trifluoromethoxy)phenyl]acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.