missing translation for 'onlineSavingsMsg'
Learn More

N1-(5-Amino-2-methylphenyl)acetamide, 97%, Thermo Scientific™

Product Code. 10746505
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10746505 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10746505 Supplier Thermo Scientific Maybridge Supplier No. BTB00549DA
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5434-30-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H12N2O
MDL Number MFCD00047859
Quantity 1 g
Synonym n-5-amino-2-methylphenyl acetamide, 2-acetylamino-4-aminotoluene, acetamide, n-5-amino-2-methylphenyl, 5'-amino-2'-methylacetanilide, 2-acetylamino-p-toluidine, n1-5-amino-2-methylphenyl acetamide, n-3-amino-6-methylphenyl acetamide, 3-acetylamino-p-toluidine, 2-methyl-5-aminoacetaniline, 3-acetamido-4-methylaniline
InChI Key UAZGSMMESOKKQZ-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)N)NC(=O)C
IUPAC Name N-(5-amino-2-methylphenyl)acetamide
Molecular Weight (g/mol) 164.208
PubChem CID 79492
Formula Weight 164.21
Percent Purity 97%
Grade Technical
Chemical Name or Material N1-(5-Amino-2-methylphenyl)acetamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.