missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ N4BP1 Recombinant Rabbit Monoclonal Antibody (24GB120)

Rabbit Recombinant Monoclonal Antibody

146.00 € - 500.00 €

Specifications

Antigen N4BP1
Clone 24GB120
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275452
View Documents
Invitrogen™
MA557109
100 μL
500.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276612
View Documents
Invitrogen™
MA557110
20 μL
146.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N4BP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Nedd4-binding partner-1 (N4BP1) is a developmentally expressed protein interactor and monoubiquitylation substrate of Nedd4, a HECT domain-containing ubiquitin ligase that mediates ubiquitylation and proteasome degradation of target proteins and may have a fundamental role to play in embryonic processes. N4BP1 can undergo Nedd4-mediated polyubiquitylation and proteosomal degradation. N4BP1 also functions as a negative regulator of AIF4 (also known as ITCH or Atrophin-1), an E3-ubiquitin ligase, by interacting with the second WW domain of AIF4 and impairing ubiquitination of substrates.
Specifications

Specifications

N4BP1
-20°C
Recombinant Monoclonal
Liquid
IgG
Mouse, Rat
N4BP1
AI481586; C81621; Kiaa0615; N4BP1; Nedd4 binding protein 1; NEDD4-binding protein 1; RGD1305179
N4BP1
Primary
Affinity chromatography
24GB120
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6A037
291921, 80750
A synthesized peptide derived from Mouse N4BP1 (aa 286-484).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.