missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ N7700 Mono-Filter, Half-Face, Air-Purifying Respirator
Description
- Product Type: Half-Face Air-Purifying Respirstor
- Filter System: Single Filter
- Cartridge Connection Type: Rd40
- Use with: Honeywell™ A2P3 Aluminum Filter Canister (12683123)
- Exhalation Valves: Twin
- Material (Body): Silicone
- Color: Black
- Size: Medium
- Harness: Four-Point, Single Unit
- Neck Strap: Yes
- Applications: Medical, Pharmaceutical, Petrochemical
Specifications
Specifications
| Cartridge/Filter | P3 |
| Certifications/Compliance (Europe) | Class 2 |
| Certifications (EN) | EN 140 |
| Connection Type | Rd40 |
| Color | Black |
| Disposable | Reusable |
| Filter Type | Single Filter |
| Material | Silicone |
| Product Type | Air-Purifying Respirator (APR) |
| Size | Medium |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?