Learn More
Invitrogen™ NA-XTD™ Influenza Neuraminidase Assay Kit
Description
Includes
Influenza Neuraminidase Assay Kit includes 1000X NA-XTD Substrate, NA Sample Prep Buffer, NA-XTD Assay Buffer, NA-XTD Accelerator and 10 detection microplates Influenza Neuraminidase Assay Reagent Set includes 1000X NA-XTD Substrate, NA Sample Prep Buffer
Next-generation chemiluminescence-based assay for long-lived emission kinetics for quantitation of influenza neuraminidase inhibitor resistance. Thermo Scientific NA-XTD Influenza Neuraminidase Assay Kit can be used for virus quantitation in media samples
The NA-XTD kit includes detection reagents and microplates, eliminating the need for luminometers equipped with a reagent injector, and improving ease-of-use. This kit also includes complete assay protocols for quantitating sensitivity to neuraminidase inhibitors in various influenza virus isolates including: human influenza types A and B, A/H1N1 pandemic, avian, equine and porcine viruses.
- Superior performance - improved, long-lived light emission kinetics and read-time flexibility
- High sensitivity - high assay signal-to-noise, detection of low-virus concentrations, and wide assay dynamic range
- Simple instrumentation requirement - no reagent injectors or luminometer needed
- Multiple applications - neuraminidase inhibitor IC50 assays and cell-based virus quantitation
Long-Lived Light Emission Kinetics and Read-Time Flexibility
The NA-XTD assay yields much longer-lived light emission kinetics than does the NA-Star assay, eliminating the need for luminometer instrumentation with reagent injectors and enabling read-time flexibility and batch-mode processing of assay plates. NA-XTD assay signal half-life is approximately 2 hours, longer than the 5-10 minute half-life of the NA-Star assay signal. IC-50 values determined from data collected immediately or up to 3 hours following addition of NA-XTD Accelerator solution are identical.
High Sensitivity
The NA-XTD chemiluminescent assay provides higher detection sensitivity (better low-end detection limit), higher assay signal-to-noise ratio, and wider assay dynamic range than fluorescent assays with the MUNANA substrate. The NA-XTD assay also typically demonstrates slightly higher signal-to-noise than the NA-Star assay. The NA-XTD assay provides 2 to 50-fold higher sensitivity by signal-to-noise ratio than MUNANA-based fluorescence assays, depending on the virus isolate. The NA-XTD assay provides a dynamic range of detection of 3 to 4 orders of magnitude of neuraminidase enzyme concentration, compared to 2 to 3 orders of magnitude range with fluorescent MUNANA assays. The wide chemiluminescent neuraminidase assay range enables determination of IC-50 values over a wide range of virus concentrations, eliminating the need to titer virus prior to performing IC50 determination assays.
Simple Instrumentation Requirement
Optimally, a small sample of culture media is removed and assayed directly with NA-XTD reagents, permitting multiple samples to be assayed over time.
Specifications
- Detection Method: Chemiluminescent
- For Use With Equipment: Luminometer (Microplate)
- Form: Liquid
- Format: 96-well plate
- High Throughput Compatibility: High Throughput-Compatible
- Readout: End Point
- Substrate: NA-XTD
- Substrate Properties: Chemical Substrate
- Substrate Type: Neuraminidase Substrate
- Target Enzyme: Neuraminidase, Sialidase
Specifications
Specifications
| Content And Storage | This kit contains 1000X NA-XTD™ Substrate, NA Sample Prep Buffer, NA-XTD™ Assay Buffer, NA-XTD™ Accelerator, and 10 detection microplates. Store all reagents at 2°C to 8°C. |
| Detection Method | Chemiluminescence |
| For Use With (Application) | Influenza Neuraminidase Assay |
| For Use With (Equipment) | Luminometer (Microplate) |
| Product Type | Neuraminidase Assay |
| Form | Liquid |
| Format | 96-well plate |
| Product Line | NA-XTD, NovaBright |
| Quantity | 10 x 96 well plates |
| Substrate | NA-XTD™ |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.