missing translation for 'onlineSavingsMsg'
Learn More

Nafcillin sodium monohydrate, MedChemExpress

Product Code. 16472229
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472229 1 g 1g
16430998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472229 Supplier Medchem Express Supplier No. HYB0555A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Nafcillin sodium monohydrate, an antibiotic, is a reversible inhibitor of β-lactamase. Nafcillin sodium monohydrate can be used for the research of staphylococcal infections.

Nafcillin sodium monohydrate, an antibiotic, is a reversible inhibitor of β-lactamase. Nafcillin sodium monohydrate can be used for the research of staphylococcal infections.

Specifications

Chemical Name or Material Nafcillin sodium monohydrate
Physical Form Solid
CAS 7177-50-6
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C21H23N2NaO6S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (220.04 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.20 mM; Need ultrasonic)
SMILES O=C([C@@H]1N(C2=O)[C@]([C@@H]2NC(C(C3=CC=CC=C3C=C4)=C4OCC)=O)([H])SC1(C)C)O[Na].O
Molecular Weight (g/mol) 454.47
Formula Weight 454.47
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.