missing translation for 'onlineSavingsMsg'
Learn More

Naftifine hydrochloride, MedChemExpress

Product Code. 16462209
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462209 1 g 1g
16400978 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462209 Supplier Medchem Express Supplier No. HYB0518A
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Naftifine hydrochloride is an antibiotic. Naftifine hydrochloride has antifungal activity against dermatophytes, aspergilli, Sporothrix schenckii, and yeasts of the genus Candida. Naftifine hydrochloride can be used for the research of superficial dermatomycoses inhibition.

Naftifine hydrochloride is an antibiotic. Naftifine hydrochloride has antifungal activity against dermatophytes, aspergilli, Sporothrix schenckii, and yeasts of the genus Candida. Naftifine hydrochloride can be used for the research of superficial dermatomycoses inhibition.

Specifications

Chemical Name or Material Naftifine hydrochloride
Physical Form Solid
CAS 65473-14-5
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C21H22ClN
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (308.78 mM; Need ultrasonic) ∣Ethanol : 25 mg/mL (77.19 mM; Need ultrasonic)
SMILES CN(C/C=C/C1=CC=CC=C1)CC2=CC=CC3=CC=CC=C23.Cl
Molecular Weight (g/mol) 323.86
Formula Weight 323.86
Percent Purity 99.38%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.