missing translation for 'onlineSavingsMsg'
Learn More

NaI @ 2.0 μg/μL, and RbI @ 0.05 μg/μl

Product Code. 30314382
Change view
Click to view available options
Quantity:
29 x 1 mL
Product Code. Quantity
30314382 29 x 1 mL
1 options
This item is not returnable. View return policy

Product Code. 30314382

Brand: VHG Labs VHGANAIRBI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rubidium iodide∣Sodium Iodide
CAS 7790-29-6, 7681-82-5
Analyte or Component Names Rubidium iodide∣Sodium Iodide
Shipping Condition Room Temperature
Certifications/Compliance ISO 9001
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solution Type Mixture
Industry Type Industrial QC
Quantity 29 x 1 mL
Concentration or Composition (by Analyte or Components) Rubidium iodide 0.05 μg/μL∣Sodium Iodide 2 μg/μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.