Learn More
Naked1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561959
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LRVKLTVAPD GSQSKRSVLV NQADLQSARP RAETKPTEDL RSWEKKQRAP LRFQGDSRLE QSGCYHHCVD ENIE Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin-Tcf signaling pathway.Specifications
Naked1 | |
Polyclonal | |
Unconjugated | |
NKD1 | |
2810434J10Rik; 9030215G15Rik; Dvl-binding protein; hNkd; hNkd1; mNkd; mNkd1; naked cuticle 1 homolog; naked cuticle 1 homolog (Drosophila); naked cuticle homolog 1; naked cuticle homolog 1 (Drosophila); naked cuticle-1; Naked1; naked-1; Nkd; NKD1; PP7246; Protein naked cuticle homolog 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85407 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969G9 | |
NKD1 | |
Recombinant protein corresponding to Human Naked1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only