missing translation for 'onlineSavingsMsg'
Learn More

Melford Nalidixic Acid Sodium Salt

Product Code. 30018576
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018576 5 g 5g
30018575 25 g 25g
30018574 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018576 Supplier Melford Supplier No. N231005.0
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nalidixic Acid Sodium Salt is a compound of notable importance in microbiology and pharmaceutical research.

Nalidixic Acid Sodium Salt is a compound of notable importance in microbiology and pharmaceutical research. This sodium salt formulation of nalidixic acid is a valuable tool for investigating bacterial resistance mechanisms and antibiotic development.

In microbiology, Nalidixic Acid Sodium Salt is widely employed to study bacterial susceptibility and resistance patterns. It serves as a selective agent in microbiological culture media, inhibiting the growth of susceptible bacteria. Researchers use it to assess the effectiveness of antibiotics and to understand the molecular mechanisms behind bacterial resistance, contributing to our understanding of microbial biology and antibiotic resistance.

Pharmaceutical researchers also rely on Nalidixic Acid Sodium Salt to develop new antibiotics and optimize existing ones. Its use as a reference standard in antimicrobial testing helps in the development of drugs that can combat emerging antibiotic-resistant bacteria, a significant challenge in modern medicine.

Specifications

Chemical Name or Material Nalidixic Acid Sodium Salt
CAS 5-8-3374
Quantity 5 g
Molecular Formula C12H11N2NaO3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.