missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sodium Hydroxide (NaOH) Solutions, 0.1–1.0 M

Product Code. 17850925
Click to view available options
Quantity:
20 L
200 L
Concentration:
0.1 M
0.5 M
1.0 M
Unit Size:
20 L
200 L
This item is not returnable. View return policy

Product Code. 17850925

Brand: Gibco™ A4782601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing.

Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing. Available in three different grades and two off-the-shelf sizes, these solutions provide outsourcing efficiencies to help reclaim time, space, and other core resources.

Features of Gibco NaOH solutions include:

  • 0.1 M, 0.5 M, and 1 M concentrations
  • Multi-compendial grade raw materials
  • cGMP manufacturing to 21CFR820
  • 20 L and 200 L ready-to-use single-use bioprocessing containers

Flexibility

Gibco NaOH solutions provide outsourcing efficiencies for customers manufacturing internally but seeking to partially outsource, as well as for customers with batch sizes that do not justify custom ordering and customers already outsourcing to Thermo Fisher Scientific but seeking additional flexibility. They are produced in the most commonly used concentrations and single-use containers.

Applications of Gibco NaOH solutions

  • pH adjustment
  • Facility/equipment sanitization
  • Chromatography column cleaning, sanitization and storage
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Melting Point 0°C
Boiling Point 100°C
Color Clear
Concentration 0.1 M
pH ≥12 to ≤14
Quantity 20 L
Molecular Formula NaOH
Packaging Bioprocess Container
Synonym Caustic soda, Acidity regulatory, Lye or Soda lye
Molecular Weight (g/mol) 40
Percent Purity 95 to 100%
Grade Compendial
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.