missing translation for 'onlineSavingsMsg'
Learn More

Naphazoline hydrochloride, MedChemExpress

Product Code. 16488488
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488488 10 g 10g
16432179 1 g 1g
16410938 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16488488 Proveedor Medchem Express N.º de proveedor HYB0446
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

MedChemExpress Naphazoline (Naphthazoline) hydrochloride is a potent α-adrenergic receptor agonist. Naphazoline hydrochloride reduces vascular hyperpermeability and promotes vasoconstriction. Naphazoline hydrochloride reduces the levels of inflammatory factors (TNF-α, IL-1β and IL-6), cytokines (IFN-γ and IL-4), IgE, GMCSF, and NGF。Naphazoline hydrochloride can be used for non-bacterial conjunctivitis research.

Naphazoline (Naphthazoline) hydrochloride is a potent α-adrenergic receptor agonist. Naphazoline hydrochloride reduces vascular hyperpermeability and promotes vasoconstriction. Naphazoline hydrochloride reduces the levels of inflammatory factors (TNF-α, IL-1β and IL-6), cytokines (IFN-γ and IL-4), IgE, GMCSF, and NGF。Naphazoline hydrochloride can be used for non-bacterial conjunctivitis research.

Especificaciones

Chemical Name or Material Naphazoline hydrochloride
Physical Form Solid
CAS 550-99-2
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C14H15ClN2
For Use With (Application) COVID-19-immunoregulation
Synonym Naphthazoline hydrochloride
Solubility Information H2O : 50 mg/mL (202.64 mM; Need ultrasonic) ∣DMSO : ≥ 25 mg/mL (101.32 mM)
SMILES C1(CC2=NCCN2)=CC=CC3=CC=CC=C13.Cl
Molecular Weight (g/mol) 246.74
Formula Weight 246.74
Percent Purity 98.37%
Grade Research
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.