missing translation for 'onlineSavingsMsg'
Learn More

Naphazoline Nitrate, TRC

Product Code. 18071910
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18071910 5 g 5g
30135418 100 g 100g
18012620 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18071910 Supplier Toronto Research Chemicals Supplier No. TRCN3439555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5144-52-5
Formula Weight 273.1113
InChI Formula InChI=1S/C14H14N2.HNO3/c1-2-7-13-11(4-1)5-3-6-12(13)10-14-15-8-9-16-14;2-1(3)4/h1-7H,8-10H2,(H,15,16);(H,2,3,4)
IUPAC Name 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;nitric acid
Molecular Formula C14 H14 N2 . H N O3
Molecular Weight (g/mol) 273.29
SMILES O[N+](=O)[O-].C(C1=NCCN1)c2cccc3ccccc23
Synonym 1H-Imidazole, 4,5-dihydro-2-(1-naphthalenylmethyl)-, nitrate (1:1), 1H-Imidazole, 4,5-dihydro-2-(1-naphthalenylmethyl)-, mononitrate (9CI), 2-Imidazoline, 2-(1-naphthylmethyl)-, mononitrate (8CI), Naftizin, Naphazoline nitrate, Naphthisin, Naphthizin, Naphthylmethyl-2-imidazoline nitrate, Naphtyzin, Privina, Privine, Privine (British), Privine nitrate, Rinazin, Sanorin
Chemical Name or Material Naphazoline nitrate

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.