accessibility menu, dialog, popup

UserName

CAS RN 5144-52-5

ONOONNH2

CAS RN 5144-52-5

IUPAC Name: 2-[(naphthalen-1-yl)methyl]-4,5-dihydro-1H-imidazol-1-ium nitrate
Synonyms: 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazol-1-ium nitrate
Molecular Weight (g/mol): 273.29
Molecular Formula: C14H15N3O3
InChi Key: HZVHPACMMGWUIV-UHFFFAOYSA-O
SMILES: [O-][N+]([O-])=O.C(C1=NCC[NH2+]1)C1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)