missing translation for 'onlineSavingsMsg'
Learn More

Naphthacene (>80%), TRC

Product Code. 30469823
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469823 1 g 1g
30466626 100 mg -
30473346 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30469823 Supplier Toronto Research Chemicals Supplier No. TRCN3776501G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-24-0
Molecular Formula C18 H12
Molecular Weight (g/mol) 228.29
Synonym 2,3-Benzanthracene, 2,3-Benzanthrene, Benz[b]anthracene, Chrysogen, Rubene, Tetracene, Tetracene (hydrocarbon)
IUPAC Name tetracene
SMILES c1ccc2cc3cc4ccccc4cc3cc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H12/c1-2-6-14-10-18-12-16-8-4-3-7-15(16)11-17(18)9-13(14)5-1/h1-12H
Formula Weight 228.0939
Chemical Name or Material Benzo(b)anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.