missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1,4-dicarboxylic acid, 98+%

Product Code. p-7048113
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11309106

Brand: Thermo Scientific Alfa Aesar L20249.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 605-70-9 | C12H8O4 | 216.19 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-70-9
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.19
MDL Number MFCD00014312
InChI Key ABMFBCRYHDZLRD-UHFFFAOYSA-N
Synonym 1,4-naphthalenedicarboxylic acid, 1,4-naphthalenedicarboxylicacid, 1,4-naphthalicacid, 1,4-naphthalic acid, pubchem21317, acmc-1b0c7, ksc357a6b, naphthalene-1,4-dicarboxylicacid, naphthalene-1,4-dicarboxlic acid, naphthaline-1,4-dicarboxylic acid
PubChem CID 69065
IUPAC Name naphthalene-1,4-dicarboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=C(C=C1)C(O)=O

Specifications

Melting Point 321°C to 325°C
Quantity 25 g
Beilstein 2051255
Solubility Information Insoluble in water.
Formula Weight 216.19
Percent Purity ≥98%
Chemical Name or Material Naphthalene-1,4-dicarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.