missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1-sulfonic acid hydrate, 98%

Product Code. 11400427
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400427 10 g 10g
11410427 50 g 50g
11420427 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400427 Supplier Thermo Scientific Alfa Aesar Supplier No. B21782.09
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6036-48-2 | C10H8O3S | 208.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6036-48-2
Molecular Formula C10H8O3S
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003987
InChI Key PSZYNBSKGUBXEH-UHFFFAOYSA-N
Synonym naphthalene-1-sulfonic acid dihydrate, 1-naphthalenesulfonic acid dihydrate, naphthalene-1-sulfonic acid hydrate
PubChem CID 71431272
IUPAC Name naphthalene-1-sulfonic acid;dihydrate
SMILES OS(=O)(=O)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 89°C to 91°C (dried)
Quantity 10 g
UN Number UN2585
Merck Index 14,6376
Formula Weight 208.23 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Naphthalene-1-sulfonic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.