missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1-sulfonic acid sodium salt, 98% (dry wt.), water <3.5%

Product Code. 11440677
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440677 100 g 100g
11420677 5 g 5g
11430677 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440677 Supplier Thermo Scientific Alfa Aesar Supplier No. B22488.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 130-14-3 | C10H7NaO3S | 230.213 g/mol

Naphthalene-1-sulfonic acid sodium salt is used as a detection reagent to investigate ion-pair high-performance liquid chromatographic retention behavior of copper(II)-1-oxa-4,7,10,13-tetraazacyclopentadecane complex. It is also used to prepare naphthalene-1-sulfonyl chloride by reaction with phosphorousoxy chloride. Further, it is used as a stationary liquid phase for resolution of dichlorophenol isomers by gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphthalene-1-sulfonic acid sodium salt is used as a detection reagent to investigate ion-pair high-performance liquid chromatographic retention behavior of copper(II)-1-oxa-4,7,10,13-tetraazacyclopentadecane complex. It is also used to prepare naphthalene-1-sulfonyl chloride by reaction with phosphorousoxy chloride. Further, it is used as a stationary liquid phase for resolution of dichlorophenol isomers by gas chromatography.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-14-3
Molecular Formula C10H7NaO3S
Molecular Weight (g/mol) 230.213
MDL Number MFCD00064964
InChI Key HIEHAIZHJZLEPQ-UHFFFAOYSA-M
Synonym sodium 1-naphthalenesulfonate, sodium naphthalene-1-sulfonate, 1-naphthalenesulfonic acid sodium salt, unii-mai7v3c3pn, sodium naphthalenesulphonate, sodium naphthalene sulfonate, 1-naphthalenesulfonic acid, sodium salt, alpha salt, sodium alpha-naphthalenesulfonate, mai7v3c3pn
PubChem CID 23661867
IUPAC Name sodium;naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 299°C to 301°C
Quantity 100 g
Beilstein 3639038
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 230.22
Percent Purity 99%
Assay (dry wt.), water <2%
Chemical Name or Material Naphthalene-1-sulfonic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.